missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Oleoyl-L-carnitine-d3 HCl (N-methyl-d3), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Oleoyl L-Carnitine-D3 (N-methyl-D3) Chloride |
| Molecular Formula | C25 D3 H45 N O4 . Cl |
| Quantity | 0.01 g |
| Synonym | (2 R)-3-Carboxy-N,N-dimethyl-N-(methyl-d3)-2-[[(9 Z)-1-oxo-9-octadecen-1-yl]oxy]-1-propanaminium chloride, Oleoyl L-carnitine-d3 chloride, Oleoyl L-carnitine-D3 (N-methyl-D3) chloride |
| InChI Formula | InChI=1 S/C25H47NO4.ClH/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-25(29)30-23(21-24(27)28)22-26(2,3)4;/h12-13,23 H,5-11,14-22H2,1-4H3;1 H/b13-12-;/t23-;/m1./s1/i2D3; |
| SMILES | [Cl-].[2 H]C([2 H])([2 H])[N+](C)(C)C[C@@H](CC(=O)O)OC(=O)CCCCCCC\C=C/CCCCCCCC |
| IUPAC Name | [(2 R)-3-carboxy-2-[(Z)-octadec-9-enoyl]oxypropyl]-dimethyl-(trideuteriomethyl)azanium;chloride |
| Molecular Weight (g/mol) | 465.124 |
| Formula Weight | 464.346 g/mol |
| Percent Purity | 99 atom % D, min 85% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction