missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(±)-Ondansetron-d5 (methylimidazole-d5), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Ondansetron-D5 (methylimidazole-D5) |
| CAS | 1219798-86-3 |
| Molecular Formula | C18 D5 H14 N3 O |
| Quantity | 0.005 g |
| Synonym | 4 H-Carbazol-4-one, 1,2,3,9-tetrahydro-9-methyl-3-[[2-(methyl-d3)-1 H-imidazol-1-yl-4,5-d2]methyl]- (ACI), 1,2,3,9-Tetrahydro-9-methyl-3-[[2-(methyl-d3)-1 H-imidazol-1-yl-4,5-d2]methyl]-4 H-carbazol-4-one (ACI), Ondansetron-d5, (±)-Ondansetron-d5 (methylimidazole-d5), Ondansetron-D5 (methylimidazole-D5) |
| InChI Formula | InChI=1 S/C18H19N3O/c1-12-19-9-10-21(12)11-13-7-8-16-17(18(13)22)14-5-3-4-6-15(14)20(16)2/h3-6,9-10,13 H,7-8,11H2,1-2H3/i1D3,9 D,10 D |
| SMILES | [2 H]c1nc(n(CC2CCc3c(C2=O)c4ccccc4n3C)c1[2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | 3-[[4,5-dideuterio-2-(trideuteriomethyl)imidazol-1-yl]methyl]-9-methyl-2,3-dihydro-1 H-carbazol-4-one |
| Molecular Weight (g/mol) | 298.394 |
| Formula Weight | 298.184 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction