missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Oxyphenbutazone-d9 (n-butyl-d9), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Oxyphenbutazone-d9 |
| CAS | 1189693-23-9 |
| Molecular Formula | C19 2H9 H11 N2 O3 |
| Quantity | 0.005 g |
| InChI Formula | InChI=1 S/C19H20N2O3/c1-2-3-9-17-18(23)20(14-7-5-4-6-8-14)21(19(17)24)15-10-12-16(22)13-11-15/h4-8,10-13,17,22 H,2-3,9H2,1H3/i1D3,2D2,3D2,9D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C1C(=O)N(N(C1=O)c2ccc(O)cc2)c3ccccc3 |
| IUPAC Name | 1-(4-hydroxyphenyl)-4-(1,1,2,2,3,3,4,4,4-nonadeuteriobutyl)-2-phenylpyrazolidine-3,5-dione |
| Molecular Weight (g/mol) | 333.43 |
| Formula Weight | 333.2039 g/mol |
| Percent Purity | 99 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction