missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(±)-p-Octopamine-alpha,beta,beta-d3 HCl, CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | rac Octopamine-d3 Hydrochloride |
| CAS | 1219803-62-9 |
| Molecular Formula | C8 2H3 H8 N O2 . Cl H |
| Quantity | 0.005 g |
| InChI Formula | InChI=1 S/C8H11NO2.ClH/c9-5-8(11)6-1-3-7(10)4-2-6;/h1-4,8,10-11 H,5,9H2;1 H/i5D2,8 D; |
| SMILES | Cl.[2 H]C([2 H])(N)C([2 H])(O)c1ccc(O)cc1 |
| IUPAC Name | 4-(2-amino-1,2,2-trideuterio-1-hydroxyethyl)phenol;hydrochloride |
| Molecular Weight (g/mol) | 192.66 |
| Formula Weight | 192.0745 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction