missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pregnenolone-2,2,3,4,4,6-d6, CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Pregnenolone-2,2,3,4,4,6-D6 |
| Molecular Formula | C21 D6 H26 O2 |
| Quantity | 0.005 g |
| Synonym | [2,2,3,4,4,6-2H6]Pregnenolone, Pregnenolone-2,2,3,4,4,6-d6, (3β)-3-Hydroxypregn-5-en-20-one-2,2,3,4,4,6-d6, Pregnenolone-2,2,3,4,4,6-D6, Pregnenolone-d6 |
| InChI Formula | InChI=1 S/C21H32O2/c1-13(22)17-6-7-18-16-5-4-14-12-15(23)8-10-20(14,2)19(16)9-11-21(17,18)3/h4,15-19,23 H,5-12H2,1-3H3/t15-,16-,17+,18-,19-,20-,21+/m0/s1/i4D,8D2,12D2,15 D |
| SMILES | [2 H]C1=C2C([2 H])([2 H])[C@@]([2 H])(O)C([2 H])([2 H])C[C@]2(C)[C@H]3 CC[C@@]4(C)[C@@H](CC[C@@H]4 C(=O)C)[C@@H]3C1 |
| IUPAC Name | 1-[(3 S,8 S,9 S,10 R,13 S,14 S,17 S)-2,2,3,4,4,6-hexadeuterio-3-hydroxy-10,13-dimethyl-1,7,8,9,11,12,14,15,16,17-decahydrocyclopenta[a]phenanthren-17-yl]ethanone |
| Molecular Weight (g/mol) | 322.5146 |
| Formula Weight | 322.2779 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction