missing translation for 'onlineSavingsMsg'
Learn More

Prometon-d3 (methoxy-d3), CDN

Product Code. 30311065
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 30311065

Brand: CDN CDND68020.01G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Prometon methoxy-d3
CAS 1219803-43-6
Molecular Formula C10 2H3 H16 N5 O
Quantity 0.01 g
Synonym Prometon D3 (methyl D3)
InChI Formula InChI=1 S/C10H19N5O/c1-6(2)11-8-13-9(12-7(3)4)15-10(14-8)16-5/h6-7 H,1-5H3,(H2,11,12,13,14,15)/i5D3
SMILES [2 H]C([2 H])([2 H])Oc1nc(NC(C)C)nc(NC(C)C)n1
IUPAC Name 2-N,4-N-di(propan-2-yl)-6-(trideuteriomethoxy)-1,3,5-triazine-2,4-diamine
Molecular Weight (g/mol) 228.31
Formula Weight 228.1778 g/mol
Percent Purity 99 atom % D, min 98% Chemical Purity
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.