missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Prometryn-d14 (di-iso-propyl-d14), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Prometryn D14 (Diisopropyl D14) |
| CAS | 1202864-57-0 |
| Molecular Formula | C10 D14 H5 N5 S |
| Quantity | 0.01 g |
| Synonym | Prometryn-d14 (di-iso-propyl-d14) |
| InChI Formula | InChI=1 S/C10H19N5S/c1-6(2)11-8-13-9(12-7(3)4)15-10(14-8)16-5/h6-7 H,1-5H3,(H2,11,12,13,14,15)/i1D3,2D3,3D3,4D3,6 D,7 D |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])(Nc1nc(NC([2 H])(C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H])nc(SC)n1)C([2 H])([2 H])[2 H] |
| IUPAC Name | 2-N,4-N-bis(1,1,1,2,3,3,3-heptadeuteriopropan-2-yl)-6-methylsulfanyl-1,3,5-triazine-2,4-diamine |
| Molecular Weight (g/mol) | 255.4426 |
| Formula Weight | 255.224 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction