missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyridaben-d13 (4-tert-butyl-d9-benzyl-2,3,5,6-d4), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Pyridaben D13 (tert-butylphenyl D13) |
| CAS | 2468638-05-1 |
| Molecular Formula | C19 D13 H12 Cl N2 O S |
| Quantity | 0.005 g |
| Synonym | Pyridaben D13 (tert-butylphenyl D13) |
| InChI Formula | InChI=1 S/C19H25ClN2OS/c1-18(2,3)14-9-7-13(8-10-14)12-24-15-11-21-22(19(4,5)6)17(23)16(15)20/h7-11 H,12H2,1-6H3/i1D3,2D3,3D3,7 D,8 D,9 D,10 D |
| SMILES | [2 H]c1c([2 H])c(c([2 H])c([2 H])c1CSC2=C(Cl)C(=O)N(N=C2)C(C)(C)C)C(C([2 H])([2 H])[2 H])(C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | 2-tert-butyl-4-chloro-5-[[2,3,5,6-tetradeuterio-4-[1,1,1,3,3,3-hexadeuterio-2-(trideuteriomethyl)propan-2-yl]phenyl]methylsulfanyl]pyridazin-3-one |
| Molecular Weight (g/mol) | 378.013 |
| Formula Weight | 377.219 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction