missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Quinaldic-d6 Acid, CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | 2-Quinolinecarboxylic Acid D6 |
| CAS | 1219802-18-2 |
| Molecular Formula | C10 D6 H N O2 |
| Quantity | 0.01 g |
| Synonym | 2-Quinolinecarboxylic-d6 acid, Quinaldic-d6 acid, 2-Carboxyquinoline-d6, Quinolin-2-carboxylic acid-d6, 2-Quinolinecarboxylic-3,4,5,6,7,8-d6 acid, 2-Quinolinecarboxylic acid D6 |
| InChI Formula | InChI=1 S/C10H7NO2/c12-10(13)9-6-5-7-3-1-2-4-8(7)11-9/h1-6 H,(H,12,13)/i1D,2 D,3 D,4 D,5 D,6 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c2c([2 H])c([2 H])c(nc2c1[2 H])C(=O)O |
| IUPAC Name | 3,4,5,6,7,8-hexadeuterioquinoline-2-carboxylic acid |
| Molecular Weight (g/mol) | 179.205 |
| Formula Weight | 179.085 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction