missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R)-(-)-Metalaxyl-d6 (2,6-dimethyl-d6), CDN
Click to view available options
Quantity:
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Metalaxyl-M D6 (dimethylphenyl D6) |
| CAS | 1398112-32-7 |
| Molecular Formula | C15 D6 H15 N O4 |
| Quantity | 0.01 g |
| Synonym | Metalaxyl-M D6 (dimethylphenyl D6), Metalaxyl-M-D6 (2,6-dimethylphenyl-D6), Metalaxyl-M-d6, N-[2,6-Di(methyl-d3)phenyl]-N-(2-methoxyacetyl)-D-alanine methyl ester, Methyl (2 R)-2-(N-(2-methoxyacetyl)-2,6-di(methyl-d3)anilino)propanoate, (2 R)-2-[N-(2-Methoxyacetyl)-2,6-bis(trideuteriomethyl)phenylamino]propanoic acid methyl ester, (2 R)-2-[N-(2-Methoxyacetyl)-2,6-bis(trideuteriomethyl)phenylamino]propanoic acid methyl ester |
| InChI Formula | InChI=1 S/C15H21NO4/c1-10-7-6-8-11(2)14(10)16(13(17)9-19-4)12(3)15(18)20-5/h6-8,12 H,9H2,1-5H3/t12-/m1/s1/i1D3,2D3 |
| SMILES | [2 H]C([2 H])([2 H])c1cccc(c1N([C@H](C)C(=O)OC)C(=O)COC)C([2 H])([2 H])[2 H] |
| IUPAC Name | methyl (2 R)-2-[N-(2-methoxyacetyl)-2,6-bis(trideuteriomethyl)anilino]propanoate |
| Molecular Weight (g/mol) | 285.3685 |
| Formula Weight | 285.1847 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction