missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Raloxifene-d10 (piperidine-d10), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Raloxifene-D10 (piperidine-D10) |
| CAS | 2512224-37-0 |
| Molecular Formula | C28 D10 H17 N O4 S |
| Quantity | 0.01 g |
| Synonym | [6-Hydroxy-2-(4-hydroxyphenyl)-1-benzothiophen-3-yl]-[4-(2-decadeuteriopiperidin-1-ylethoxy)phenyl]methanone, 2-(4-Hydroxyphenyl)-3-[4-[2-(piperidin-1-yl-d10)ethoxy-d4]benzoyl]-1-benzothiophen-6-ol, [6-Hydroxy-2-(4-hydroxyphenyl)-1-benzothiophen-3-yl]-[4-(2-piperidin-d10-1-ylethoxy)phenyl]methanone, Raloxifene-d10, Raloxifene-D10 (piperidine-D10) |
| InChI Formula | InChI=1 S/C28H27NO4S/c30-21-8-4-20(5-9-21)28-26(24-13-10-22(31)18-25(24)34-28)27(32)19-6-11-23(12-7-19)33-17-16-29-14-2-1-3-15-29/h4-13,18,30-31 H,1-3,14-17H2/i1D2,2D2,3D2,14D2,15D2 |
| SMILES | [2 H]C1([2 H])N(CCOc2ccc(cc2)C(=O)c3c(sc4cc(O)ccc34)c5ccc(O)cc5)C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C1([2 H])[2 H] |
| IUPAC Name | [4-[2-(2,2,3,3,4,4,5,5,6,6-decadeuteriopiperidin-1-yl)ethoxy]phenyl]-[6-hydroxy-2-(4-hydroxyphenyl)-1-benzothiophen-3-yl]methanone |
| Molecular Weight (g/mol) | 483.645 |
| Formula Weight | 483.229 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction