missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S-Sulfo-DL-cysteine-2,3,3-d3, CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | S-Sulfo-DL-cysteine-2,3,3-D3 |
| CAS | 2687960-80-9 |
| Molecular Formula | C3 D3 H4 N O5 S2 |
| Quantity | 0.005 g |
| Synonym | 2-Amino-3-(sulfosulfanyl)propanoic-2,3,3-d3 acid, S-Sulfo-DL-cysteine-2,3,3-d3, S-Sulfo-DL-cysteine-2,3,3-D3 |
| InChI Formula | InChI=1 S/C3H7NO5S2/c4-2(3(5)6)1-10-11(7,8)9/h2H,1,4H2,(H,5,6)(H,7,8,9)/i1D2,2 D |
| SMILES | [2 H]C([2 H])(SS(=O)(=O)O)C([2 H])(N)C(=O)O |
| IUPAC Name | 2-amino-2,3,3-trideuterio-3-sulfosulfanylpropanoic acid |
| Molecular Weight (g/mol) | 204.24 |
| Formula Weight | 203.995 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction