missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Cyclamate-d11 (cyclohexyl-d11), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Cyclamic Acid-d11 Sodium Salt |
| Molecular Formula | C6 D11 H N O3 S . Na |
| Quantity | 0.01 g |
| Synonym | Cyclohexylsulfamic Acid-d11 Monosodium Salt, Acofarinas-d11, Assugrin-d11, Asugryn-d11, Azucrona-d11, Cyclamate Sodium-d11, Cyclohexylsulfamate Sodium-d11, Cyclohexylsulfamic Acid Sodium Salt-d11, Dulzor-Etas-d11, Duplo-Sacarin-d11, Hachi-Sugar-d11, Ibiosuc-d11, N-Cyclohexylsulfamic Acid-d11 Sodium Salt, Natreen-d11, Nutter-d11, Sodium Cyclamate-d11, Sodium cyclohexanesulfamate-d11, Sodium Cyclohexylaminesulfonate-d11, Sodium Cyclohexylsulfamate-d11, Sodium Sucaryl-d11, Sucaryl Sodium-d11, Sucrum 7-d11, Suessette-d11, Suestamin-d11, Sugarin-d11, Sugaron-d11, Sodium Cyclamate D11 (Cyclohexyl D11), (1,2,2,3,3,4,4,5,5,6,6-Undecadeuteriocyclohexyl)sulfamic acid sodium salt, Sodium cyclamate-D11 (cyclohexyl-D11), Cyclamic-d11 acid sodium salt, N-(Cyclohexyl-d11)sulfamic acid sodium salt, Cyclamate-d11 sodium, [2H11]Cyclohexylamidosulfuric acid sodium salt, Cyclohexyl-d11-aminesulfonic acid sodium salt, Cyclohexylsulfamic acid-d11 sodium salt |
| InChI Formula | InChI=1 S/C6H13NO3S.Na/c8-11(9,10)7-6-4-2-1-3-5-6;/h6-7 H,1-5H2,(H,8,9,10);/q;+1/p-1/i1D2,2D2,3D2,4D2,5D2,6 D; |
| SMILES | [Na+].[2 H]C1([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])(NS(=O)(=O)[O-])C([2 H])([2 H])C1([2 H])[2 H] |
| IUPAC Name | sodium;N-(1,2,2,3,3,4,4,5,5,6,6-undecadeuteriocyclohexyl)sulfamate |
| Molecular Weight (g/mol) | 212.29 |
| Formula Weight | 212.1126 g/mol |
| Percent Purity | 99 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction