missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfadiazine-d4 (benzene-d4), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Sulfadiazine D4 |
| CAS | 1020719-78-1 |
| Molecular Formula | C10 D4 H6 N4 O2 S |
| Quantity | 0.005 g |
| Synonym | 4-Amino-N-2-pyrimidinylbenzene-2,3,5,6-d4-sulfonamide, Benzene-2,3,5,6-d4-sulfonamide, 4-amino-N-2-pyrimidinyl-, Sulfadiazine D4 |
| InChI Formula | InChI=1 S/C10H10N4O2S/c11-8-2-4-9(5-3-8)17(15,16)14-10-12-6-1-7-13-10/h1-7 H,11H2,(H,12,13,14)/i2D,3 D,4 D,5 D |
| SMILES | [2 H]c1c([2 H])c(c([2 H])c([2 H])c1N)S(=O)(=O)Nc2ncccn2 |
| IUPAC Name | 4-amino-2,3,5,6-tetradeuterio-N-pyrimidin-2-ylbenzenesulfonamide |
| Molecular Weight (g/mol) | 254.302 |
| Formula Weight | 254.078 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction