missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfadimethoxine-d6 (dimethoxy-d6), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Sulfadimethoxine D6 (2,6-dimethoxy D6) |
| CAS | 73068-02-7 |
| Molecular Formula | C12 2H6 H8 N4 O4 S |
| Quantity | 0.01 g |
| Synonym | Sulfadimethoxine D6 (2,6-dimethoxy D6) |
| InChI Formula | InChI=1 S/C12H14N4O4S/c1-19-11-7-10(14-12(15-11)20-2)16-21(17,18)9-5-3-8(13)4-6-9/h3-7 H,13H2,1-2H3,(H,14,15,16)/i1D3,2D3 |
| SMILES | [2 H]C([2 H])([2 H])Oc1cc(NS(=O)(=O)c2ccc(N)cc2)nc(OC([2 H])([2 H])[2 H])n1 |
| IUPAC Name | 4-amino-N-[2,6-bis(trideuteriomethoxy)pyrimidin-4-yl]benzenesulfonamide |
| Molecular Weight (g/mol) | 316.37 |
| Formula Weight | 316.1112 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction