missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Testosterone-16,16,17-d3 3-Phenylpropionate, CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Testosterone-16,16,17-d3 3-Phenylpropionate |
| CAS | 876054-55-6 |
| Molecular Formula | C28 2H3 H33 O3 |
| Quantity | 0.01 g |
| Synonym | Androst-4-en-3-one-16,16,17-d3, 17-(1-oxo-3-phenylpropoxy)-, (17β)- (9 CI), Testosterone-16,16,17-d3 17-phenylpropionate, Testosterone-16,16,17-d3 17-Phenylpropionate |
| InChI Formula | InChI=1 S/C28H36O3/c1-27-16-14-21(29)18-20(27)9-10-22-23-11-12-25(28(23,2)17-15-24(22)27)31-26(30)13-8-19-6-4-3-5-7-19/h3-7,18,22-25 H,8-17H2,1-2H3/t22-,23-,24-,25-,27-,28-/m0/s1/i12D2,25 D |
| SMILES | C[C@@]12 [C@@]3([C@]([C@]4([C@](C)(CC3)[C@@](OC(CCC5=CC=CC=C5)=O)(C(C4)([2 H])[2 H])[2 H])[H])(CCC1=CC(=O)CC2)[H])[H] |
| IUPAC Name | [(8 R,9 S,10 R,13 S,14 S,17 S)-16,16,17-trideuterio-10,13-dimethyl-3-oxo-1,2,6,7,8,9,11,12,14,15-decahydrocyclopenta[a]phenanthren-17-yl] 3-phenylpropanoate |
| Molecular Weight (g/mol) | 423.6 |
| Formula Weight | 423.2853 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction