missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Testosterone-16,16,17-d3 Acetate, CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Testosterone-16,16,17-D3 Acetate |
| Molecular Formula | C21 D3 H27 O3 |
| Quantity | 0.005 g |
| Synonym | 16,16,17-Trideuterio-4-androsten-17β-ol-3-one 17-acetate, 17-O-Acetyltestosterone-16,16,17-d3, (17β)-17-Acetoxyandrost-4-ene-3-one-16,16,17-d3, Testosterone-16,16,17-d3 17-acetate, Testosterone-16,16,17-d3 acetate, Testosterone-16,16,17-D3 acetate |
| InChI Formula | InChI=1 S/C21H30O3/c1-13(22)24-19-7-6-17-16-5-4-14-12-15(23)8-10-20(14,2)18(16)9-11-21(17,19)3/h12,16-19 H,4-11H2,1-3H3/t16-,17-,18-,19-,20-,21-/m0/s1/i7D2,19 D |
| SMILES | [2 H]C1([2 H])C[C@H]2 [C@@H]3 CCC4=CC(=O)CC[C@]4(C)[C@H]3 CC[C@]2(C)[C@@]1([2 H])OC(=O)C |
| IUPAC Name | [(8 R,9 S,10 R,13 S,14 S,17 S)-16,16,17-trideuterio-10,13-dimethyl-3-oxo-1,2,6,7,8,9,11,12,14,15-decahydrocyclopenta[a]phenanthren-17-yl] acetate |
| Molecular Weight (g/mol) | 333.48 |
| Formula Weight | 333.238 g/mol |
| Percent Purity | 99 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction