missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetramethyl-d12-thiuram Disulfide, CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | ThiramD12 |
| CAS | 69193-86-8 |
| Molecular Formula | C6 2H12 N2 S4 |
| Quantity | 0.01 g |
| Synonym | Thiram D12 |
| InChI Formula | InChI=1 S/C6H12N2S4/c1-7(2)5(9)11-12-6(10)8(3)4/h1-4H3/i1D3,2D3,3D3,4D3 |
| SMILES | [2 H]C([2 H])([2 H])N(C(=S)SSC(=S)N(C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | bis(trideuteriomethyl)carbamothioylsulfanyl N,N-bis(trideuteriomethyl)carbamodithioate |
| Molecular Weight (g/mol) | 252.51 |
| Formula Weight | 252.0637 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction