missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thiacloprid-d4 (thiazolidinylidene-d4), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Thiacloprid ethylene-d4 |
| CAS | 1793071-39-2 |
| Molecular Formula | C10 2H4 H5 Cl N4 S |
| Quantity | 0.005 g |
| Synonym | Thiacloprid-d4, [N(Z)]-N-[3-[(6-chloro-3-pyridinyl)methyl]-2-thiazolidinylidene]cyanamide-d4, [N(Z)]-[3-[(6-Chloro-3-pyridinyl)methyl]-2-thiazolidinylidene]cyanamide, Bariard-d4, Biscaya, Calypso 70 WG-d4, Calypso 70 WG-d4, Calypso 70 WG-d4, Calypso 70 WG-d4 |
| InChI Formula | InChI=1 S/C10H9ClN4S/c11-9-2-1-8(5-13-9)6-15-3-4-16-10(15)14-7-12/h1-2,5 H,3-4,6H2/b14-10-/i3D2,4D2 |
| SMILES | [2 H]C1([2 H])S\C(=N/C#N)\N(Cc2ccc(Cl)nc2)C1([2 H])[2 H] |
| IUPAC Name | (Z)-[3-[(6-chloropyridin-3-yl)methyl]-4,4,5,5-tetradeuterio-1,3-thiazolidin-2-ylidene]cyanamide |
| Molecular Weight (g/mol) | 256.75 |
| Formula Weight | 256.0488 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction