missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tolfenamic-d4 Acid (benzoic-3,4,5,6-d4), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Tolfenamic Acid-d4 |
| CAS | 1246820-82-5 |
| Molecular Formula | C14 D4 H8 Cl N O2 |
| Quantity | 0.01 g |
| Synonym | 2-[(3-Chloro-2-methylphenyl)amino]-3,4,5,6-tetradeuteriobenzoic acid, Tolfenamic-d4 Acid (benzoic-3,4,5,6-d4), Tolfenamic Acid-d4, Tolfenamic Acid-D4 (benzoic-3,4,5,6-D4), 2-[(3-Chloro-2-methylphenyl)amino]benzoic-3,4,5,6-d4 acid, N-(2-Methyl-3-chlorophenyl)anthranilic acid-3,4,5,6-d4 |
| InChI Formula | InChI=1 S/C14H12ClNO2/c1-9-11(15)6-4-8-12(9)16-13-7-3-2-5-10(13)14(17)18/h2-8,16 H,1H3,(H,17,18)/i2D,3 D,5 D,7 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c(C(=O)O)c(Nc2cccc(Cl)c2C)c1[2 H] |
| IUPAC Name | 2-(3-chloro-2-methylanilino)-3,4,5,6-tetradeuteriobenzoic acid |
| Molecular Weight (g/mol) | 265.728 |
| Formula Weight | 265.081 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction