missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tris(2-chloroethyl) Phosphate-d12, CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | Tris(2-chloroethyl)phosphate-D12 |
| CAS | 1276500-47-0 |
| Molecular Formula | C6 2H12 Cl3 O4 P |
| Quantity | 0.01 g |
| InChI Formula | InChI=1 S/C6H12Cl3O4P/c7-1-4-11-14(10,12-5-2-8)13-6-3-9/h1-6H2/i1D2,2D2,3D2,4D2,5D2,6D2 |
| SMILES | [2 H]C([2 H])(Cl)C([2 H])([2 H])OP(=O)(OC([2 H])([2 H])C([2 H])([2 H])Cl)OC([2 H])([2 H])C([2 H])([2 H])Cl |
| IUPAC Name | tris(2-chloro-1,1,2,2-tetradeuterioethyl) phosphate |
| Molecular Weight (g/mol) | 297.56 |
| Formula Weight | 296.0292 g/mol |
| Percent Purity | 99 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction