missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(±)-Voriconazole-d3 (difluorophenyl-d3), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | (+/-)-Voriconazole-D3 (difluorophenyl-D3) |
| Molecular Formula | C16 D3 H11 F3 N5 O |
| Quantity | 0.01 g |
| Synonym | (αR,βS)-rel-α-(2,4-Difluorophenyl-3,5,6-d3)-5-fluoro-β-methyl-α-(1 H-1,2,4-triazol-1-ylmethyl)-4-pyrimidineethanol, (R*,S*)-α-(2,4-Difluorophenyl-d3)-5-fluoro-β-methyl-α-(1 H-1,2,4-triazol-1-ylmethyl)-4-pyrimidineethanol, (±)-Voriconazole-d3, (±)-Voriconazole-D3 (difluorophenyl-D3), (2 RS,3 SR)-2-(2,4-Difluorophenyl-3,5,6-d3)-3-(5-fluoropyrimidin-4-yl)-1-(1 H-1,2,4-triazol-1-yl)butan-2-ol |
| InChI Formula | InChI=1 S/C16H14F3N5O/c1-10(15-14(19)5-20-7-22-15)16(25,6-24-9-21-8-23-24)12-3-2-11(17)4-13(12)18/h2-5,7-10,25 H,6H2,1H3/t10-,16+/m0/s1/i2D,3 D,4 D |
| SMILES | [2 H]c1c([2 H])c(c(F)c([2 H])c1F)[C@@](O)(Cn2cncn2)[C@@H](C)c3ncncc3F |
| IUPAC Name | (2 R,3 S)-3-(5-fluoropyrimidin-4-yl)-1-(1,2,4-triazol-1-yl)-2-(2,3,5-trideuterio-4,6-difluorophenyl)butan-2-ol |
| Molecular Weight (g/mol) | 352.329 |
| Formula Weight | 352.134 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction