missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Bromoeicosane-d41, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | 1-Bromoeicosane-D41 |
| CAS | 202480-71-5 |
| Molecular Formula | C20 D41 Br |
| Quantity | 0.05 g |
| Synonym | Eicosane-1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,13,13,14,14,15,15,16,16,17,17,18,18,19,19,20,20-d41, 20-bromo- (9 CI), 20-Bromoeicosane-1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,13,13,14,14,15,15,16,16,17,17,18,18,19,19,20,20-d41, 20-Bromoeicosane-d41, 1-Bromoeicosane-d41, 1-Bromoeicosane-D41, Eicosyl-d41 bromide |
| InChI Formula | InChI=1 S/C20H41Br/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21/h2-20H2,1H3/i1D3,2D2,3D2,4D2,5D2,6D2,7D2,8D2,9D2,10D2,11D2,12D2,13D2,14D2,15D2,16D2,17D2,18D2,19D2,20D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])Br |
| IUPAC Name | 1-bromo-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,13,13,14,14,15,15,16,16,17,17,18,18,19,19,20,20,20-hentetracontadeuterioicosane |
| Molecular Weight (g/mol) | 402.6962 |
| Formula Weight | 401.4965 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction