missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(±)-1-Hexadecanoyl-3-chloropropane-d5-diol, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | 3-Chloro-1,2-propanediol 1-Palmitate d5 |
| CAS | 1189718-15-7 |
| Molecular Formula | C19 2H5 H32 Cl O3 |
| Quantity | 0.05 g |
| Synonym | 3-Chloro-1,2-propanediol 1-Palmitate D5 |
| InChI Formula | InChI=1 S/C19H37ClO3/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-19(22)23-17-18(21)16-20/h18,21 H,2-17H2,1H3/i16D2,17D2,18 D |
| SMILES | [2 H]C([2 H])(Cl)C([2 H])(O)C([2 H])([2 H])OC(=O)CCCCCCCCCCCCCCC |
| IUPAC Name | (3-chloro-1,1,2,3,3-pentadeuterio-2-hydroxypropyl) hexadecanoate |
| Molecular Weight (g/mol) | 353.98 |
| Formula Weight | 353.2745 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction