missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2-Bis(hydroxymethyl-d2)propionic-3,3,3-d3 Acid, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.1g
Specifications
Specifications
| Chemical Name or Material | 2,2-Dimethylolpropionic Acid-D7 |
| CAS | 1219802-03-5 |
| Molecular Formula | C5 D7 H3 O4 |
| Quantity | 0.05 g |
| Synonym | Propanoic-3,3,3-d3 acid, 2,2-bis(hydroxymethyl-d2)- (ACI), 2,2-Bis(hydroxymethyl-d2)propanoic-3,3,3-d3 acid (ACI), 2,2-Bis(hydroxymethyl-d2)propionic-3,3,3-d3 Acid, 2,2-Dimethylolpropionic Acid-D7 |
| InChI Formula | InChI=1 S/C5H10O4/c1-5(2-6,3-7)4(8)9/h6-7 H,2-3H2,1H3,(H,8,9)/i1D3,2D2,3D2 |
| SMILES | [2 H]C([2 H])([2 H])C(C(=O)O)(C([2 H])([2 H])O)C([2 H])([2 H])O |
| IUPAC Name | 3,3,3-trideuterio-2,2-bis[dideuterio(hydroxy)methyl]propanoic acid |
| Molecular Weight (g/mol) | 141.1736 |
| Formula Weight | 141.1018 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction