missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(3,4-Dihydroxyphenyl)ethyl-1-13C-amine HCl, CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | Dopamine-13 C (2-aminoethyl-2-13 C) Hydrochloride |
| CAS | 335081-04-4 |
| Molecular Formula | 13 C C7 H11 N O2 . Cl H |
| Quantity | 0.05 g |
| Synonym | 1,2-Benzenediol, 4-(2-aminoethyl-2-13 C)-, hydrochloride (9 CI), Dopamine-13 C hydrochloride, Dopamine-13 C (2-aminoethyl-2-13 C) hydrochloride, 4-(2-Aminoethyl-2-13 C)-1,2-benzenediol hydrochloride, 2-(3,4-Dihydroxyphenyl)ethyl-1-13 C-amine HCl |
| InChI Formula | InChI=1 S/C8H11NO2.ClH/c9-4-3-6-1-2-7(10)8(11)5-6;/h1-2,5,10-11 H,3-4,9H2;1 H/i4+1; |
| SMILES | Cl.N[13 CH2]Cc1ccc(O)c(O)c1 |
| IUPAC Name | 4-(2-amino(213 C)ethyl)benzene-1,2-diol;hydrochloride |
| Molecular Weight (g/mol) | 190.632 |
| Formula Weight | 190.059 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction