missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(3,4-Dimethoxy-d6-phenyl)propionic-2,2,3,3-d4 Acid, CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Specifications
Specifications
| Chemical Name or Material | 3-(3,4-Dimethoxyphenyl)propanoic acid dimethoxy,ethyl-d10 |
| CAS | 1398065-72-9 |
| Molecular Formula | C11 2H10 H4 O4 |
| Quantity | 0.05 g |
| Synonym | 3-(3,4-Dimethoxy-d6-phenyl)propionic-2,2,3,3-d4 acid, 3-(3,4-Dimethoxyphenyl)propanoic acid D10 (dimethoxy-D6,ethyl-D4), 3,4-Di(methoxy-d3)benzenepropanoic-2,2,3,3-d4 acid |
| InChI Formula | InChI=1 S/C11H14O4/c1-14-9-5-3-8(4-6-11(12)13)7-10(9)15-2/h3,5,7 H,4,6H2,1-2H3,(H,12,13)/i1D3,2D3,4D2,6D2 |
| SMILES | [2 H]C([2 H])([2 H])Oc1ccc(cc1OC([2 H])([2 H])[2 H])C([2 H])([2 H])C([2 H])([2 H])C(=O)O |
| IUPAC Name | 3-[3,4-bis(trideuteriomethoxy)phenyl]-2,2,3,3-tetradeuteriopropanoic acid |
| Molecular Weight (g/mol) | 220.29 |
| Formula Weight | 220.152 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction