missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,5-Dibromo-4-hydroxybenzonitrile-2,6-d2, CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | Bromoxynil-D2 |
| CAS | 1219798-95-4 |
| Molecular Formula | C7 D2 H Br2 N O |
| Quantity | 0.05 g |
| Synonym | 3,5-Dibromo-4-hydroxybenzonitrile-2,6-d2, 1-Cyano-3,5-dibromo-4-hydroxybenzene-2,6-d2, 2,6-Dibromo-4-cyanophenol-3,5-d2, Bromoxynil-d2, Bromoxynil-D2 |
| InChI Formula | InChI=1 S/C7H3Br2NO/c8-5-1-4(3-10)2-6(9)7(5)11/h1-2,11 H/i1D,2 D |
| SMILES | [2 H]c1c(Br)c(O)c(Br)c([2 H])c1C#N |
| IUPAC Name | 3,5-dibromo-2,6-dideuterio-4-hydroxybenzonitrile |
| Molecular Weight (g/mol) | 278.9251 |
| Formula Weight | 276.8707 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction