missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(2,4-Dichlorophenoxy-d3)butyric Acid, CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | 2,4-DB D3 (phenyl-D3) |
| CAS | 1219802-46-6 |
| Molecular Formula | C10 D3 H7 Cl2 O3 |
| Quantity | 0.05 g |
| Synonym | 4-(2,4-Dichlorophenoxy-d3)butyric Acid, 2,4-DB-d3 (ring-d3), 2,4-DB D3 (phenyl-D3), 4-(2,4-Dichlorophenoxy-3,5,6-d3)butanoic acid |
| InChI Formula | InChI=1 S/C10H10Cl2O3/c11-7-3-4-9(8(12)6-7)15-5-1-2-10(13)14/h3-4,6 H,1-2,5H2,(H,13,14)/i3D,4 D,6 D |
| SMILES | [2 H]c1c([2 H])c(OCCCC(=O)O)c(Cl)c([2 H])c1Cl |
| IUPAC Name | 4-(2,4-dichloro-3,5,6-trideuteriophenoxy)butanoic acid |
| Molecular Weight (g/mol) | 252.1091 |
| Formula Weight | 251.0195 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction