missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Cholesten-3-one-2,2,4,6,6-d5, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | 4-Cholesten-3-one-2,2,4,6,6-d5 |
| CAS | 72560-60-2 |
| Molecular Formula | C27 D5 H39 O |
| Quantity | 0.05 g |
| Synonym | Cholest-4-en-3-one-2,2,4,6,6-d5 (9 CI), 4-Cholesten-3-one-2,2,4,6,6-d5, 4-Cholesten-3-one-2,2,4,6,6-D5, delta;4-Cholesten-3-one-d5, (17β)-17-Octylandrost-4-en-3-one-2,2,4,6,6-d5 |
| InChI Formula | InChI=1 S/C27H44O/c1-18(2)7-6-8-19(3)23-11-12-24-22-10-9-20-17-21(28)13-15-26(20,4)25(22)14-16-27(23,24)5/h17-19,22-25 H,6-16H2,1-5H3/t19-,22+,23-,24+,25+,26+,27-/m1/s1/i9D2,13D2,17 D |
| SMILES | [2 H]C1=C2C([2 H])([2 H])C[C@H]3 [C@@H]4 CC[C@H]([C@H](C)CCCC(C)C)[C@@]4(C)CC[C@@H]3 [C@@]2(C)CC([2 H])([2 H])C1=O |
| IUPAC Name | (8 S,9 S,10 R,13 R,14 S,17 R)-2,2,4,6,6-pentadeuterio-10,13-dimethyl-17-[(2 R)-6-methylheptan-2-yl]-1,7,8,9,11,12,14,15,16,17-decahydrocyclopenta[a]phenanthren-3-one |
| Molecular Weight (g/mol) | 389.668 |
| Formula Weight | 389.371 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction