missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Piperidine-d9-carboxamide, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | Isonipecotamide-D9 |
| CAS | 1219803-20-9 |
| Molecular Formula | C6 D9 H3 N2 O |
| Quantity | 0.05 g |
| Synonym | 4-Piperidine-2,2,3,3,4,5,5,6,6-d9-carboxamide (ACI), 4-Piperidine-d9-carboxamide, 4-Piperidinecarboxamide-d9, 4-Carbamoylpiperidine-d9, 4-Carboxamidopiperidine-2,2,3,3,4,5,5,6,6-d9, 4-Piperidylcarboxamide-d9, Isonipecotamide-D9 |
| InChI Formula | InChI=1 S/C6H12N2O/c7-6(9)5-1-3-8-4-2-5/h5,8 H,1-4H2,(H2,7,9)/i1D2,2D2,3D2,4D2,5 D |
| SMILES | [2 H]C1([2 H])NC([2 H])([2 H])C([2 H])([2 H])C([2 H])(C(=O)N)C1([2 H])[2 H] |
| IUPAC Name | 2,2,3,3,4,5,5,6,6-nonadeuteriopiperidine-4-carboxamide |
| Molecular Weight (g/mol) | 137.2277 |
| Formula Weight | 137.1515 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction