missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-(Hydroxymethyl-d2)uracil-6-d1, CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | 5-Hydroxymethyluracil-D3 |
| CAS | 352438-73-4 |
| Molecular Formula | C5 H3 D3 N2 O3 |
| Quantity | 0.05 g |
| Synonym | 2,4(1 H,3 H)-Pyrimidinedione-6-d, 5-(hydroxymethyl-d2)- (9 CI), 5-(Hydroxymethyl-d2)-2,4(1 H,3 H)-pyrimidinedione-6-d, 5-(Hydroxymethyl-d2)uracil-6-d1, α-Hydroxythymine-d3, 5-Hydroxymethyluracil-D3 |
| InChI Formula | InChI=1 S/C5H6N2O3/c8-2-3-1-6-5(10)7-4(3)9/h1,8 H,2H2,(H2,6,7,9,10)/i1D,2D2 |
| SMILES | O=C1C(C([2 H])([2 H])O)=C([2 H])NC(N1)=O |
| IUPAC Name | 6-deuterio-5-[dideuterio(hydroxy)methyl]-1 H-pyrimidine-2,4-dione |
| Molecular Weight (g/mol) | 145.1312 |
| Formula Weight | 145.0567 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction