missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis(2-chloroethyl)-d8 Ether, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | Bis(2-chloroethyl)ether-d8 |
| CAS | 93952-02-4 |
| Molecular Formula | C4 D8 Cl2 O |
| Quantity | 0.05 g |
| Synonym | Ethane-1,1,2,2-d4, 1,1'-oxybis[2-chloro- (9 CI), Bis(2-chloroethyl-1,1,2,2-d4) Ether, Bis(2-chloroethyl) Ether-d8, Bis(2-chloroethyl) Ether-D8, 1,1'-Oxybis[2-chloroethane-1,1,2,2-d4], β,β'-Dichlorodiethyl ether-d8, β,β'-Dichlorodiethyl ether-d8 |
| InChI Formula | InChI=1 S/C4H8Cl2O/c5-1-3-7-4-2-6/h1-4H2/i1D2,2D2,3D2,4D2 |
| SMILES | [2 H]C([2 H])(Cl)C([2 H])([2 H])OC([2 H])([2 H])C([2 H])([2 H])Cl |
| IUPAC Name | 1-chloro-2-(2-chloro-1,1,2,2-tetradeuterioethoxy)-1,1,2,2-tetradeuterioethane |
| Molecular Weight (g/mol) | 151.061 |
| Formula Weight | 150.0454 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction