missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis(2-ethoxyethyl) Phthalate-3,4,5,6-d4, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.10g
Specifications
Specifications
| Chemical Name or Material | Phthalic acid, bis-2-ethoxyethyl ester D4 |
| CAS | 1398066-12-0 |
| Molecular Formula | C16 2H4 H18 O6 |
| Quantity | 0.05 g |
| InChI Formula | InChI=1 S/C16H22O6/c1-3-19-9-11-21-15(17)13-7-5-6-8-14(13)16(18)22-12-10-20-4-2/h5-8 H,3-4,9-12H2,1-2H3/i5D,6 D,7 D,8 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c(C(=O)OCCOCC)c(C(=O)OCCOCC)c1[2 H] |
| IUPAC Name | bis(2-ethoxyethyl) 3,4,5,6-tetradeuteriobenzene-1,2-dicarboxylate |
| Molecular Weight (g/mol) | 314.37 |
| Formula Weight | 314.1667 g/mol |
| Percent Purity | 99 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction