missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cholesterol-3,4-13C2, CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | Cholesterol 3,4-13C2 |
| CAS | 78887-48-6 |
| Molecular Formula | 13C2 C25 H46 O |
| Quantity | 0.05 g |
| Synonym | Cholest-5-en-3-ol-3,4-13C2, (3β)- (9 CI, ACI), (3β)-Cholest-5-en-3-ol-3,4-13C2 (ACI), [3,4-13C2]-Cholesterol, Cholesterol-3,4-13C2, Cholesterol-13C2 |
| InChI Formula | InChI=1 S/C27H46O/c1-18(2)7-6-8-19(3)23-11-12-24-22-10-9-20-17-21(28)13-15-26(20,4)25(22)14-16-27(23,24)5/h9,18-19,21-25,28 H,6-8,10-17H2,1-5H3/t19-,21+,22+,23-,24+,25+,26+,27-/m1/s1/i17+1,21+1 |
| SMILES | CC(C)CCC[C@@H](C)[C@H]1 CC[C@H]2 [C@@H]3 CC=C4[13 CH2][13 C@@H](O)CC[C@]4(C)[C@H]3 CC[C@]12 C |
| IUPAC Name | (3 S,8 S,9 S,10 R,13 R,14 S,17 R)-10,13-dimethyl-17-[(2 R)-6-methylheptan-2-yl]-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1 H-cyclopenta[a]phenanthren-3-ol |
| Molecular Weight (g/mol) | 388.6389 |
| Formula Weight | 388.3616 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction