missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cholic Acid-24-13C, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | Cholic Acid-24-13 C |
| CAS | 52886-36-9 |
| Molecular Formula | 13 C C23 H40 O5 |
| Quantity | 0.05 g |
| Synonym | Cholan-24-oic-24-13 C acid, 3,7,12-trihydroxy-, (3α,5β,7α,12α)- (9 CI, ACI), (3α,5β,7α,12α)-3,7,12-Trihydroxycholan-24-oic-24-13 C acid (ACI), Cholic-24-13 C acid, Cholic Acid-24-13 C |
| InChI Formula | InChI=1 S/C24H40O5/c1-13(4-7-21(28)29)16-5-6-17-22-18(12-20(27)24(16,17)3)23(2)9-8-15(25)10-14(23)11-19(22)26/h13-20,22,25-27 H,4-12H2,1-3H3,(H,28,29)/t13-,14+,15-,16-,17+,18+,19-,20+,22+,23+,24-/m1/s1/i21+1 |
| SMILES | C[C@H](CC[13 C](=O)O)[C@H]1 CC[C@H]2 [C@@H]3 [C@H](O)C[C@@H]4 C[C@H](O)CC[C@]4(C)[C@H]3 C[C@H](O)[C@]12 C |
| IUPAC Name | (4 R)-4-[(3 R,5 S,7 R,8 R,9 S,10 S,12 S,13 R,14 S,17 R)-3,7,12-trihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1 H-cyclopenta[a]phenanthren-17-yl](113 C)pentanoic acid |
| Molecular Weight (g/mol) | 409.564 |
| Formula Weight | 409.291 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction