missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diphenhydramine-d5 HCl (phenyl-d5), CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | Diphenhydramine-D5 (phenyl-D5) Hydrochloride |
| CAS | 1219795-16-0 |
| Molecular Formula | C17H16D5NO • HCl |
| Quantity | 0.05 g |
| Synonym | Diphenhydramine-d5 Hydrochloride (Phenyl-d5);2-[Phenyl(phenyl-d5)methoxy]-N,N-dimethylethanamine hydrochloride, Diphenhydramine-d5 hydrochloride, Diphenhydramine-D5 (phenyl-D5) hydrochloride |
| InChI Formula | InChI=1 S/C17H21NO/c1-18(2)13-14-19-17(15-9-5-3-6-10-15)16-11-7-4-8-12-16/h3-12,17 H,13-14H2,1-2H3/i3D,5 D,6 D,9 D,10 D |
| SMILES | CN(C)CCOC(C1=CC=CC=C1)C(C([2 H])=C2[2 H])=C(C([2 H])=C2[2 H])[2 H].Cl |
| IUPAC Name | N,N-dimethyl-2-(phenyl(phenyl-d5)methoxy)ethan-1-amine hydrochloride |
| Molecular Weight (g/mol) | 296.85 |
| Formula Weight | 296.17 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction