missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Disodium Etidronate-d3 (methyl-d3), CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | Etidronate methyl-d3 disodium salt |
| CAS | 358730-93-5 |
| Molecular Formula | C2 2H3 H3 O7 P2 . 2 Na |
| Quantity | 0.05 g |
| Synonym | Phosphonic acid, (1-hydroxyethylidene-2,2,2-d3)bis-, disodium salt (9 CI), Etidronic Acid-d3 Sodium Salt, Etidronate-D3 Disodium (methyl D3) |
| InChI Formula | InChI=1 S/C2H8O7P2.2 Na/c1-2(3,10(4,5)6)11(7,8)9;;/h3H,1H3,(H2,4,5,6)(H2,7,8,9);;/q;2*+1/p-2/i1D3;; |
| SMILES | [Na+].[Na+].[2 H]C([2 H])([2 H])C(O)(P(=O)(O)[O-])P(=O)(O)[O-] |
| IUPAC Name | disodium;hydroxy-[2,2,2-trideuterio-1-hydroxy-1-[hydroxy(oxido)phosphoryl]ethyl]phosphinate |
| Molecular Weight (g/mol) | 253.01 |
| Formula Weight | 252.9572 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction