missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexadecanoic-11,11,12,12-d4 Acid, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | Hexadecanoic-11,11,12,12-D4 Acid |
| CAS | 75736-57-1 |
| Molecular Formula | C16 D4 H28 O2 |
| Quantity | 0.05 g |
| Synonym | Hexadecanoic-11,11,12,12-d4 acid (9 CI), Hexadecanoic-11,11,12,12-D4 Acid, Palmitic acid-11,11,12,12-d4, [11,11,12,12-2H4]Hexadecanoic acid, [11,11,12,12-2H4]Palmitic acid, Hexadecanoic Acid-11,11,12,12-d4 |
| InChI Formula | InChI=1 S/C16H32O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16(17)18/h2-15H2,1H3,(H,17,18)/i5D2,6D2 |
| SMILES | [2 H]C([2 H])(CCCC)C([2 H])([2 H])CCCCCCCCCC(=O)O |
| IUPAC Name | 11,11,12,12-tetradeuteriohexadecanoic acid |
| Molecular Weight (g/mol) | 260.4487 |
| Formula Weight | 260.2653 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction