missing translation for 'onlineSavingsMsg'
Learn More
Learn More
iso-Butyl 4-Hydroxybenzoate-2,3,5,6-d4, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | 4-Hydroxybenzoic acid-isobutyl ester ring-d4 |
| CAS | 1219805-33-0 |
| Molecular Formula | C11 2H4 H10 O3 |
| Quantity | 0.05 g |
| Synonym | 4-Hydroxybenzoic acid-isobutyl ester D4 (ring D4) |
| InChI Formula | InChI=1 S/C11H14O3/c1-8(2)7-14-11(13)9-3-5-10(12)6-4-9/h3-6,8,12 H,7H2,1-2H3/i3D,4 D,5 D,6 D |
| SMILES | [2 H]c1c([2 H])c(C(=O)OCC(C)C)c([2 H])c([2 H])c1O |
| IUPAC Name | 2-methylpropyl 2,3,5,6-tetradeuterio-4-hydroxybenzoate |
| Molecular Weight (g/mol) | 198.25 |
| Formula Weight | 198.1194 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction