missing translation for 'onlineSavingsMsg'
Learn More
Learn More
mono-iso-Propyl Phthalate-3,4,5,6-d4, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | Monoisopropyl Phthalate-d4 |
| CAS | 2733160-00-2 |
| Molecular Formula | C11 D4 H8 O4 |
| Quantity | 0.05 g |
| Synonym | mono-iso-Propyl Phthalate-3,4,5,6-d4, Monoisopropyl Phthalate-d4, Phthalic Acid Monoisopropyl Ester-d4, Phthalic Acid-d4 Monoisopropyl Ester, Monoisopropyl Phthalate-3,4,5,6-D4 |
| InChI Formula | InChI=1 S/C11H12O4/c1-7(2)15-11(14)9-6-4-3-5-8(9)10(12)13/h3-7 H,1-2H3,(H,12,13)/i3D,4 D,5 D,6 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c(C(=O)OC(C)C)c(C(=O)O)c1[2 H] |
| IUPAC Name | 2,3,4,5-tetradeuterio-6-propan-2-yloxycarbonylbenzoic acid |
| Molecular Weight (g/mol) | 212.2352 |
| Formula Weight | 212.0987 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction