missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Acetyl-S-benzyl-2,3,4,5,6-d5-DL-cysteine, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | N-Acetyl-S-benzyl-2,3,4,5,6-d5-DL-cysteine |
| Molecular Formula | C12H10D5NO3S |
| Quantity | 0.05 g |
| Synonym | N-Acetyl-S-(phenylmethyl)cysteine-d5, N-Acetyl-S-benzyl-DL-cysteine-d5; |
| InChI Formula | InChI=1 S/C12H15NO3S/c1-9(14)13-11(12(15)16)8-17-7-10-5-3-2-4-6-10/h2-6,11 H,7-8H2,1H3,(H,13,14)(H,15,16)/i2D,3 D,4 D,5 D,6 D |
| SMILES | [2 H]C1=C(C([2 H])=C([2 H])C([2 H])=C1[2 H])CSCC(NC(C)=O)C(O)=O |
| IUPAC Name | N-acetyl-S-((phenyl-d5)methyl)cysteine |
| Molecular Weight (g/mol) | 258.35 |
| Formula Weight | 258.11 g/mol |
| Percent Purity | 99 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction