missing translation for 'onlineSavingsMsg'
Learn More
Learn More
n-Decyltrimethylammonium-d30 Bromide, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.10g
Specifications
Specifications
| Chemical Name or Material | Decyltrimethylammonium-D30 Bromide |
| Molecular Formula | C13 D30 N . Br |
| Quantity | 0.05 g |
| Synonym | N,N,N-Tri(methyl-d3)-1-decan-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-d21-aminium bromide, Decyl-d21-tri(methyl-d3)ammonium bromide, Decyltrimethylammonium-D30 bromide, Decyltrimethylammonium-d30 bromide, N,N,N-Tri(methyl-d3)-1-decan-d21-aminium bromide, N-Henicosadeuteriodecyl-N,N,N-tris(trideuteriomethyl)ammonium bromide, n-Decyltrimethylammonium-d30 bromide |
| InChI Formula | InChI=1 S/C13H30N.BrH/c1-5-6-7-8-9-10-11-12-13-14(2,3)4;/h5-13H2,1-4H3;1 H/q+1;/p-1/i1D3,2D3,3D3,4D3,5D2,6D2,7D2,8D2,9D2,10D2,11D2,12D2,13D2; |
| SMILES | [Br-].[2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])[N+](C([2 H])([2 H])[2 H])(C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | 1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-henicosadeuteriodecyl-tris(trideuteriomethyl)azanium;bromide |
| Molecular Weight (g/mol) | 310.473 |
| Formula Weight | 309.344 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction