missing translation for 'onlineSavingsMsg'
Learn More
Learn More
n-Dodecylphosphocholine-d38, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | Dodecylphosphocholine-D38 |
| CAS | 130890-78-7 |
| Molecular Formula | C17 D38 N O4 P |
| Quantity | 0.05 g |
| Synonym | Ethan-1,1,2,2-d4-aminium, 2-[[(dodecyl-d25-oxy)hydroxyphosphinyl]oxy]-N,N,N-tri(methyl-d3)-, inner salt (9 CI, ACI), n-Dodecylphosphocholine-d38, 2-[[(Dodecyl-d25-oxy)hydroxyphosphinyl]oxy]-N,N,N-tri(methyl-d3)ethan-1,1,2,2-d4-aminium inner salt |
| InChI Formula | InChI=1 S/C17H38NO4P/c1-5-6-7-8-9-10-11-12-13-14-16-21-23(19,20)22-17-15-18(2,3)4/h5-17H2,1-4H3/i1D3,2D3,3D3,4D3,5D2,6D2,7D2,8D2,9D2,10D2,11D2,12D2,13D2,14D2,15D2,16D2,17D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])OP(=O)([O-])OC([2 H])([2 H])C([2 H])([2 H])[N+](C([2 H])([2 H])[2 H])(C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | 1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,12-pentacosadeuteriododecyl [1,1,2,2-tetradeuterio-2-[tris(trideuteriomethyl)azaniumyl]ethyl] phosphate |
| Molecular Weight (g/mol) | 389.6958 |
| Formula Weight | 389.4924 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction