missing translation for 'onlineSavingsMsg'
Learn More
Learn More
n-Hexadecyl-d33-amine, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | Hexadecylamine-D33 |
| CAS | 1191245-82-5 |
| Molecular Formula | C16 D33 H2 N |
| Quantity | 0.05 g |
| Synonym | 1-Hexadecan-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,13,13,14,14,15,15,16,16,16-d33-amine (ACI), Hexadecylamine-D33, 1-Hexadecylamine-D33, Hexadecylamine-d33, 1-Aminohexadecane-d33, Cetylamine-d33, n-Hexadecyl-d33-amine |
| InChI Formula | InChI=1 S/C16H35N/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17/h2-17H2,1H3/i1D3,2D2,3D2,4D2,5D2,6D2,7D2,8D2,9D2,10D2,11D2,12D2,13D2,14D2,15D2,16D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])N |
| IUPAC Name | 1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,13,13,14,14,15,15,16,16,16-tritriacontadeuteriohexadecan-1-amine |
| Molecular Weight (g/mol) | 274.659 |
| Formula Weight | 274.484 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction