missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenylbutazone-d10 (diphenyl-d10), CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | Phenylbutazone diphenyl-d10 |
| CAS | 1219794-69-0 |
| Molecular Formula | C19 2H10 H10 N2 O2 |
| Quantity | 0.05 g |
| Synonym | 4-Butyl-1,2-bis(pentadeuteriophenyl)pyrazolidine-3,5-dione, 4-Butyl-1,2-di(phenyl-d5)pyrazolidine-3,5-dione, Phenylbutazone (ring D10), Phenylbutazone (diphenyl-D10), Phenylbutazone-d10 |
| InChI Formula | InChI=1 S/C19H20N2O2/c1-2-3-14-17-18(22)20(15-10-6-4-7-11-15)21(19(17)23)16-12-8-5-9-13-16/h4-13,17 H,2-3,14H2,1H3/i4D,5 D,6 D,7 D,8 D,9 D,10 D,11 D,12 D,13 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c(N2N(C(=O)C(CCCC)C2=O)c3c([2 H])c([2 H])c([2 H])c([2 H])c3[2 H])c([2 H])c1[2 H] |
| IUPAC Name | 4-butyl-1,2-bis(2,3,4,5,6-pentadeuteriophenyl)pyrazolidine-3,5-dione |
| Molecular Weight (g/mol) | 318.44 |
| Formula Weight | 318.2152 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction