missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Serotonin-alpha,alpha,beta,beta-d4 Creatinine Sulfate Complex H2O, CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | Serotonin-D4 (2-aminoethyl-1,1,2,2-D4) Creatinine Sulfate Monohydrate |
| CAS | 2687960-56-9 |
| Molecular Formula | C10 D4 H8 N2 O . C4 H7 N3 O . H2 O4 S . H2 O |
| Quantity | 0.05 g |
| Synonym | 4 H-Imidazol-4-one, 2-amino-1,5-dihydro-1-methyl-, compd. with 3-(2-aminoethyl-1,1,2,2-d4)-1 H-indol-5-ol sulfate, hydrate (1:1:1:1) (ACI), 5-Hydroxytryptamine-d4 creatinine sulfate monohydrate, Serotonin-d4 creatinine sulfate monohydrate, Serotonin-alpha,alpha,beta,beta-d4 Creatinine Sulfate Complex H2O |
| InChI Formula | InChI=1 S/C10H12N2O.C4H7N3O.H2O4S.H2O/c11-4-3-7-6-12-10-2-1-8(13)5-9(7)10;1-7-2-3(8)6-4(7)5;1-5(2,3)4;/h1-2,5-6,12-13 H,3-4,11H2;2H2,1H3,(H2,5,6,8);(H2,1,2,3,4);1H2/i3D2,4D2;;; |
| SMILES | O.[2 H]C([2 H])(N)C([2 H])([2 H])c1c[nH]c2ccc(O)cc12.CN3CC(=O)NC3=N.OS(=O)(=O)O |
| IUPAC Name | 3-(2-amino-1,1,2,2-tetradeuterioethyl)-1 H-indol-5-ol;2-imino-1-methylimidazolidin-4-one;sulfuric acid;hydrate |
| Molecular Weight (g/mol) | 409.4514 |
| Formula Weight | 409.1569 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction