missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium (±)-3-Hydroxybutyrate-2,2-d2, CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | rac 3-Hydroxybutyric acid 2,2-d2 sodium salt |
| CAS | 352439-00-0 |
| Molecular Formula | C4 2H2 H5 O3 . Na |
| Quantity | 0.05 g |
| InChI Formula | InChI=1 S/C4H8O3.Na/c1-3(5)2-4(6)7;/h3,5 H,2H2,1H3,(H,6,7);/q;+1/p-1/i2D2; |
| SMILES | [Na+].[2 H]C([2 H])(C(C)O)C(=O)[O-] |
| IUPAC Name | sodium;2,2-dideuterio-3-hydroxybutanoate |
| Molecular Weight (g/mol) | 128.1 |
| Formula Weight | 128.0418 g/mol |
| Percent Purity | 99 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction