missing translation for 'onlineSavingsMsg'
Learn More
Learn More
trans-4-Hydroxy-L-proline-2,5,5-d3, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | (4 R)-4-Hydroxy-L-proline-2,5,5-D3 |
| CAS | 1356016-86-8 |
| Molecular Formula | C5 D3 H6 N O3 |
| Quantity | 0.05 g |
| Synonym | L-Proline-2,5,5-d3, 4-hydroxy-, (4 R)-, trans-4-Hydroxy-L-proline-2,5,5-D3, (2 S,4 R)-4-Hydroxyproline-2,5,5-d3, (2 S,4 R)-4-Hydroxy-2,5,5-trideuteriopyrrolidine-2-carboxylic acid |
| InChI Formula | InChI=1 S/C5H9NO3/c7-3-1-4(5(8)9)6-2-3/h3-4,6-7 H,1-2H2,(H,8,9)/t3-,4+/m1/s1/i2D2,4 D |
| SMILES | [2 H]C1([2 H])N[C@@]([2 H])(C[C@H]1 O)C(=O)O |
| IUPAC Name | (2 S,4 R)-2,5,5-trideuterio-4-hydroxypyrrolidine-2-carboxylic acid |
| Molecular Weight (g/mol) | 134.1483 |
| Formula Weight | 134.0771 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction