missing translation for 'onlineSavingsMsg'
Learn More

0.115 M (0.115 N) Sodium Hydroxide Low in Carbonate Analytical Volumetric Solution (AVL), Reagecon
BIOPHARMA

Product Code. 16451174 Shop All Reagecon Products
Change view
Click to view available options
0.115 M (0.115 N) Sodium Hydroxide Low in Carbonate Analytical Volumetric Solution (AVL), 1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
16451174 0.115 M (0.115 N) Sodium Hydroxide Low in Carbonate Analytical Volumetric Solution (AVL), 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16451174 Supplier Reagecon Supplier No. S21151LC

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Fully factorized, high purity, stable product developed and tested for acid/base titrations

  • Tested to ± 0.1%
  • May be used for manual titrations or more commonly automatic titrations

Specifications

Concentration 0.115 M
Quantity 1 L
Packaging Amber glass Winchester bottle GL45
Physical Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.